Merck
  • Culturing INS-1 cells on CDPGYIGSR-, RGD- and fibronectin surfaces improves insulin secretion and cell proliferation.
Culturing INS-1 cells on CDPGYIGSR-, RGD- and fibronectin surfaces improves insulin secretion and cell proliferation.
Acta biomaterialia (2011-11-17)
Carina Kuehn, Evan A Dubiel, Georges Sabra, Patrick Vermette
ABSTRACT
Rat insulinoma cells (INS-1), an immortalized pancreatic beta cell line, were cultured on low-fouling carboxymethyl-dextran (CMD) layers bearing fibronectin, the tripeptide Arg-Gly-Asp (RGD) or CDPGYIGSR, a laminin nonapeptide. INS-1 cells were non-adherent on CMD and RGE but adhered to fibronectin- and peptide-coated CMD surfaces and to tissue culture polystyrene (TCPS). On CMD bearing fibronectin and the peptides, INS-1 cells showed higher glucose-stimulated insulin secretion compared to those on TCPS, bare CMD and RGE. INS-1 cells experienced a net cell growth, with the lowest found after 7 days on CMD and the highest on fibronectin. Similarly, cells on RGD and CDPGYIGSR showed lower net growth rates than those on fibronectin. Expression of E-cadherin and integrins αvβ3 and α5 were similar between the conditions, except for α5 expression on fibronectin, RGD and CDPGYIGSR. Larger numbers of Ki-67-positive cells were found on CDPGYIGSR, TCPS, fibronectin and RGD. Cells in all conditions expressed Pdx1.
Sigma-Aldrich
CM-Dextran sodium salt, BioXtra