Merck
  • Comparison of Microneutralization and Histo-Blood Group Antigen-Blocking Assays for Functional Norovirus Antibody Detection.
Comparison of Microneutralization and Histo-Blood Group Antigen-Blocking Assays for Functional Norovirus Antibody Detection.
The Journal of infectious diseases (2019-10-16)
Robert L Atmar, Khalil Ettayebi, B Vijayalakshmi Ayyar, Frederick H Neill, Ralph P Braun, Sasirekha Ramani, Mary K Estes
ABSTRACT
The development of an in vitro cultivation system for human noroviruses allows the measurement of neutralizing antibody levels. Serum neutralizing antibody levels were determined using a GII.4/Sydney/2012-like virus in human intestinal enteroids in samples collected before and 4 weeks after administration of an investigational norovirus vaccine and were compared with those measured in histo-blood group antigen (HBGA)-blocking assays. Neutralizing antibody seroresponses were observed in 71% of 24 vaccinated adults, and antibody levels were highly correlated (r = 0.82, P < .001) with those measured by HBGA blocking. HBGA-blocking antibodies are a surrogate for neutralization in human noroviruses. NCT02475278.
Sigma-Aldrich
Sodium glycochenodeoxycholate