Merck
  • Bioorganometallic derivatives of 4-hydrazino-benzenesulphonamide as carbonic anhydrase inhibitors: synthesis, characterisation and biological evaluation.
Bioorganometallic derivatives of 4-hydrazino-benzenesulphonamide as carbonic anhydrase inhibitors: synthesis, characterisation and biological evaluation.
Journal of enzyme inhibition and medicinal chemistry (2020-02-11)
Jeremie Brichet, Rodrigo Arancibia, Emanuela Berrino, Claudiu T Supuran
ABSTRACT
A series of bio-organometallic-hydrazones of the general formula [{(η5-C5H4)-C(R)=N-N(H)-C6H4-4-SO2NH2}]MLn(MLn = Re(CO)3, Mn(CO)3, FeCp; R=H, CH3) were prepared by reaction of formyl/acetyl organometallic precursors with 4-hydrazino-benzenesulphonamide. All compounds were characterized by conventional spectroscopic techniques (infra-red, 1H and 13C NMR, mass spectrometry and elemental analysis). Biological evaluation as carbonic anhydrase (CA, EC 4.2.1.1) inhibitors agents was carried out using four human/h) isoforms, hCA I, II, IX and XII. The cytosolic isoforms hCA I and II were effectively inhibited by almost all derivatives with inhibition constants of 1.7-22.4 nM. Similar effects were observed for the tumour-associated transmembrane isoform hCA XII (KIs of 1.9-24.4 nM). hCA IX was less sensitive to inhibition with these compounds. The presence of bio-organometallic or metallo-carbonyl moieties in the molecules of these CAIs makes them amenable for interesting pharmacologic applications, for example for compounds with CO donating properties.
Sigma-Aldrich
Acetylferrocene, 95%