Merck
  • A molecular modeling study of the interaction of 2'-fluoro-substituted analogues of dUMP/FdUMP with thymidylate synthase.
A molecular modeling study of the interaction of 2'-fluoro-substituted analogues of dUMP/FdUMP with thymidylate synthase.
Bioorganic & medicinal chemistry letters (2008-03-26)
Adam Jarmuła, Anna Dowierciał, Wojciech Rode
ABSTRACT
Molecular dynamics simulations and free energy calculations are presented, exploring previously described experimentally studied interactions of a series of 2'-fluoro-substituted dUMP/FdUMP analogues with thymidylate synthase (TS). The results show the inhibitory behaviors of 2'-F-ara-UMP, 2',2''-diF-dUMP and 2',5-diF-ara-UMP to be dependent upon the binding positions and orientations adopted by the molecules of these compounds in the active site of TS. The binding mode of 2',5-diF-ara-UMP suggests a novel role of the active site residue Trp 80, stabilizing through hydrophobic stacking the binding position of the pyrimidine ring in 2',5-diF-ara-UMP.
Sigma-Aldrich
5-Fluoro-2′-deoxyuridine 5′-monophosphate sodium salt, ~85%