Merck
  • Thiol-Ene Clickable Gelatin: A Platform Bioink for Multiple 3D Biofabrication Technologies.
Thiol-Ene Clickable Gelatin: A Platform Bioink for Multiple 3D Biofabrication Technologies.
Advanced materials (Deerfield Beach, Fla.) (2017-10-19)
Sarah Bertlein, Gabriella Brown, Khoon S Lim, Tomasz Jungst, Thomas Boeck, Torsten Blunk, Joerg Tessmar, Gary J Hooper, Tim B F Woodfield, Juergen Groll
ABSTRACT
Bioprinting can be defined as the art of combining materials and cells to fabricate designed, hierarchical 3D hybrid constructs. Suitable materials, so called bioinks, have to comply with challenging rheological processing demands and rapidly form a stable hydrogel postprinting in a cytocompatible manner. Gelatin is often adopted for this purpose, usually modified with (meth-)acryloyl functionalities for postfabrication curing by free radical photopolymerization, resulting in a hydrogel that is cross-linked via nondegradable polymer chains of uncontrolled length. The application of allylated gelatin (GelAGE) as a thiol-ene clickable bioink for distinct biofabrication applications is reported. Curing of this system occurs via dimerization and yields a network with flexible properties that offer a wider biofabrication window than (meth-)acryloyl chemistry, and without additional nondegradable components. An in-depth analysis of GelAGE synthesis is conducted, and standard UV-initiation is further compared with a recently described visible-light-initiator system for GelAGE hydrogel formation. It is demonstrated that GelAGE may serve as a platform bioink for several biofabrication technologies by fabricating constructs with high shape fidelity via lithography-based (digital light processing) 3D printing and extrusion-based 3D bioprinting, the latter supporting long-term viability postprinting of encapsulated chondrocytes.
Sigma-Aldrich
Ruthenium photoinitiator kit