Merck
  • Single molecule characterization of individual extracellular vesicles from pancreatic cancer.
Single molecule characterization of individual extracellular vesicles from pancreatic cancer.
Journal of extracellular vesicles (2019-11-20)
Kathleen M Lennon, Devin L Wakefield, Adam L Maddox, Matthew S Brehove, Ari N Willner, Krystine Garcia-Mansfield, Bessie Meechoovet, Rebecca Reiman, Elizabeth Hutchins, Marcia M Miller, Ajay Goel, Patrick Pirrotte, Kendall Van Keuren-Jensen, Tijana Jovanovic-Talisman
ABSTRACT
Biofluid-accessible extracellular vesicles (EVs) may represent a new means to improve the sensitivity and specificity of detecting disease. However, current methods to isolate EVs encounter challenges when they are used to select specific populations. Moreover, it has been difficult to comprehensively characterize heterogeneous EV populations at the single vesicle level. Here, we robustly assessed heterogeneous EV populations from cultured cell lines via nanoparticle tracking analysis, proteomics, transcriptomics, transmission electron microscopy, and quantitative single molecule localization microscopy (qSMLM). Using qSMLM, we quantified the size and biomarker content of individual EVs. We applied qSMLM to patient plasma samples and identified a pancreatic cancer-enriched EV population. Our goal is to advance single molecule characterization of EVs for early disease detection. Abbreviations: EV: Extracellular Vesicle; qSMLM: quantitative Single Molecule Localization Microscopy; PDAC: Pancreatic Ductal Adenocarcinoma; EGFR: epidermal growth factor receptor 1; CA19-9: carbohydrate antigen 19-9; SEC: size exclusion chromatography; WGA: wheat germ agglutinin; AF647: Alexa Fluor 647; Ab: antibody; HPDEC: Healthy Pancreatic Ductal Epithelial Cell; TEM: Transmission Electron Microscopy.
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, 2 mg/mL, Chemicon®