Merck
  • Hu1D10 induces apoptosis concurrent with activation of the AKT survival pathway in human chronic lymphocytic leukemia cells.
Hu1D10 induces apoptosis concurrent with activation of the AKT survival pathway in human chronic lymphocytic leukemia cells.
Blood (2003-11-25)
Andrew P Mone, Peng Huang, Helene Pelicano, Carolyn M Cheney, Jennifer M Green, J Yun Tso, Amy J Johnson, Sara Jefferson, Thomas S Lin, John C Byrd
ABSTRACT
The 1D10 antigen is the target for Hu1D10 (apolizumab), a humanized HLA-DR beta-chain-specific antibody that is currently in clinical trials for hematologic malignancies. We demonstrate that Hu1D10 induces caspase-independent apoptosis following secondary cross-linking in primary chronic lymphocytic leukemia (CLL) cells. Generation of reactive oxygen species (ROS) and signal transduction, as evidenced by phosphorylation of Syk and AKT, were noted. The source of the Hu1D10-induced ROS was examined using the Raji lymphoblastic cell line with engineered defects in the mitochondrial respiratory chain. Hu1D10 treatment of clones with deficient mitochondrial respiration produced ROS suggesting a cytoplasmic source. Administration of ROS scavengers to primary CLL cells prior to Hu1D10 treatment diminished AKT activation. Treatment with Hu1D10 and the phosphatidylinositol 3-kinase inhibitor LY294002 demonstrated in vitro synergy with enhanced apoptosis. In conjunction with an ongoing clinical trial, blood samples were collected following intravenous infusion of Hu1D10 and analyzed for phosphorylation of AKT. Two of 3 patient samples showed a sustained increase in AKT phosphorylation following Hu1D10 administration. These data suggest that Hu1D10 ligation in CLL cells induces death and survival signals for which combination therapies may be designed to greatly enhance efficiency of both Hu1D10 and other class II antibodies in development.
Sigma-Aldrich
Anti-ZAP-70 Antibody, clone 2F3.2, clone 2F3.2, Upstate®, from mouse
Sigma-Aldrich
Piceatannol, A cell-permeable, substrate competitive and reversible plant metabolite that inhibits the activity of rat liver protein kinase A catalytic subunit (IC₅₀ = 3 µM), PKC (IC₅₀ = 8 µM), MLCK (IC₅₀ = 12 µM).
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
LY 294002, LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC₅₀ = 1.4 µM). Acts on the ATP-binding site.
Sigma-Aldrich
Caspase Inhibitor VI, Z-VAD-FMK, CAS 161401-82-7, is an irreversible pan caspase inhibitor. Does not require pretreatment with esterase for in vitro studies.