Merck
  • First report of sylvatic DENV-2-associated dengue hemorrhagic fever in West Africa.
First report of sylvatic DENV-2-associated dengue hemorrhagic fever in West Africa.
PLoS neglected tropical diseases (2011-08-11)
Leticia Franco, Gustavo Palacios, José Antonio Martinez, Ana Vázquez, Nazir Savji, Fernando De Ory, María Paz Sanchez-Seco, Dolores Martín, W Ian Lipkin, Antonio Tenorio
ABSTRACT
Dengue virus (DENV) circulates in human and sylvatic cycles. Sylvatic strains are both ecologically and evolutionarily distinct from endemic viruses. Although sylvatic dengue cycles occur in West African countries and Malaysia, only a few cases of mild human disease caused by sylvatic strains and one single case of dengue hemorrhagic fever in Malaysia have been reported. Here we report a case of dengue hemorrhagic fever (DHF) with thrombocytopenia (13000/µl), a raised hematocrit (32% above baseline) and mucosal bleeding in a 27-year-old male returning to Spain in November 2009 after visiting his home country Guinea Bissau. Sylvatic DENV-2 West African lineage was isolated from blood and sera. This is the first case of DHF associated with sylvatic DENV-2 in Africa and the second case worldwide of DHF caused by a sylvatic strain.
Sigma-Aldrich
Anti-Dengue Virus Type II Antibody, clone 3H5-1, clone 3H5-1, Chemicon®, from mouse