Merck
  • Deficiency of liver-derived insulin-like growth factor-I (IGF-I) does not interfere with the skin wound healing rate.
Deficiency of liver-derived insulin-like growth factor-I (IGF-I) does not interfere with the skin wound healing rate.
PloS one (2018-03-14)
Ileana Ruxandra Botusan, Xiaowei Zheng, Sampath Narayanan, Jacob Grünler, Vivekananda Gupta Sunkari, Freja S Calissendorff, Ishrath Ansurudeen, Christopher Illies, Johan Svensson, John-Olov Jansson, Claes Ohlsson, Kerstin Brismar, Sergiu-Bogdan Catrina
ABSTRACT
IGF-I is a growth factor, which is expressed in virtually all tissues. The circulating IGF-I is however derived mainly from the liver. IGF-I promotes wound healing and its levels are decreased in wounds with low regenerative potential such as diabetic wounds. However, the contribution of circulating IGF-I to wound healing is unknown. Here we investigated the role of systemic IGF-I on wound healing rate in mice with deficiency of liver-derived IGF-I (LI-IGF-I-/- mice) during normal (normoglycemic) and impaired wound healing (diabetes). LI-IGF-I-/- mice with complete inactivation of the IGF-I gene in the hepatocytes were generated using the Cre/loxP recombination system. This resulted in a 75% reduction of circulating IGF-I. Diabetes was induced with streptozocin in both LI-IGF-I-/- and control mice. Wounds were made on the dorsum of the mice, and the wound healing rate and histology were evaluated. Serum IGF-I and GH were measured by RIA and ELISA respectively. The expression of IGF-I, IGF-II and the IGF-I receptor in the skin were evaluated by qRT-PCR. The local IGF-I protein expression in different cell types of the wounds during wound healing process was analyzed using immunohistochemistry. The wound healing rate was similar in LI-IGF-I-/- mice to that in controls. Diabetes significantly delayed the wound healing rate in both LI-IGF-I-/- and control mice. However, no significant difference was observed between diabetic animals with normal or reduced hepatic IGF-I production. The gene expression of IGF-I, IGF-II and IGF-I receptor in skin was not different between any group of animals tested. Local IGF-I levels in the wounds were similar between of LI-IGF-I-/- and WT mice although a transient reduction of IGF-I expression in leukocytes in the wounds of LI-IGF-I-/- was observed seven days post wounding. Deficiency in the liver-derived IGF-I does not affect wound healing in mice, neither in normoglycemic conditions nor in diabetes.
Sigma-Aldrich
Anti-CD11b Antibody (human/mouse), APC-Cy7, clone M1/70, clone M1/70, 0.2 mg/mL, from rat