Merck
  • Live-cell imaging of alkyne-tagged small biomolecules by stimulated Raman scattering.
Live-cell imaging of alkyne-tagged small biomolecules by stimulated Raman scattering.
Nature methods (2014-03-04)
Lu Wei, Fanghao Hu, Yihui Shen, Zhixing Chen, Yong Yu, Chih-Chun Lin, Meng C Wang, Wei Min
ABSTRACT
Sensitive and specific visualization of small biomolecules in living systems is highly challenging. We report stimulated Raman-scattering imaging of alkyne tags as a general strategy for studying a broad spectrum of small biomolecules in live cells and animals. We demonstrate this technique by tracking alkyne-bearing drugs in mouse tissues and visualizing de novo synthesis of DNA, RNA, proteins, phospholipids and triglycerides through metabolic incorporation of alkyne-tagged small precursors.
Sigma-Aldrich
17-Octadecynoic acid, ≥95% (GC)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)