Merck
Studies on the stereoselectivity of the P2-purinoceptor.
British journal of pharmacology (1983-08-01)
G Burnstock, N J Cusack, J M Hills, I MacKenzie, P Meghji
ABSTRACT
ATP, 2-chloro-ATP, 2-methylthio-ATP, and their unnatural L-enantiomers, were synthesized and their effects tested on the guinea-pig taenia coli and urinary bladder, and the stimulated frog ventricle. The potent P2-purinoceptor agonists, 2-chloro-ATP and 2-methylthio-ATP were, respectively, 30 and 200 times more effective than ATP in relaxing the guinea-pig taenia, but approximately as effective as ATP in contracting the guinea-pig bladder and augmenting the force of contraction of the frog ventricle. A high degree of stereoselectivity was observed for relaxations of the guinea-pig taenia coli produced by the P2-purinoceptoragonists, and 2-methylthio-ATP was over 700 times more effective than its L-enantiomer. In contrast, stereoselectivity for contraction of the guinea-pig bladder was observed only at low concentrations with each pair of enantiomers, and a similar low stereoselectivity was displayed by the frog ventricle. These results show that P2-purinoceptors mediating inhibitory responses in the guinea-pig taenia coli can show a high degree of stereoselectivity, while P2-purinoceptors mediating excitatory responses in the guinea-pig bladder and in the frog ventricle show little stereoselectivity. The partial stereoselectivity of the P2-purinoceptor in smooth muscle contrasts with the absolute stereospecificity of P1-purinoceptors for adenosine on smooth muscle and autonomic nerve terminals and the absolute stereospecificity of the receptor for ADP on the human platelet.