Merck
  • Fibroblasts from patients with idiopathic pulmonary fibrosis are resistant to cisplatin-induced cell death via enhanced CK2-dependent XRCC1 activity.
Fibroblasts from patients with idiopathic pulmonary fibrosis are resistant to cisplatin-induced cell death via enhanced CK2-dependent XRCC1 activity.
Apoptosis : an international journal on programmed cell death (2019-03-10)
Jintaek Im, Richard Seonghun Nho
ABSTRACT
Idiopathic pulmonary fibrosis (IPF) is a deadly and progressive fibrotic lung disease, but the precise etiology remains elusive. IPF is characterized by the presence of apoptosis-resistant (myo)fibroblasts that relentlessly produce a collagen-rich extracellular matrix (ECM). Recent studies showed that an anti-cancer chemotherapy drug cisplatin is implicated in the development of pulmonary fibrosis, suggesting that the treatment of cancer patients with cisplatin may alter fibroblast viability. To address this possibility, we investigated the cisplatin-induced cell death mechanism in lung fibroblasts derived from IPF and non-IPF patients in response to a collagen matrix. IPF fibroblasts showed enhanced resistance to cisplatin-induced cell death compared to non-IPF fibroblasts in a time- and dose-dependent manner. Molecular study showed that the expression of γH2AX, PUMA and caspase-3/7 activity was abnormally reduced in IPF fibroblasts, suggesting that DNA damage-induced apoptosis caused by cisplatin was suppressed in IPF fibroblasts. Our study further revealed that DNA repair protein XRCC1 activity was aberrantly increased as a result of CK2 hyper-activation in cisplatin-treated IPF fibroblasts, and this alteration protected IPF fibroblasts from cisplatin-induced cell death. Our results showed that IPF fibroblasts residing in a collagen rich matrix are resistance to cisplatin-induced cell death due to the aberrantly high CK2/XRCC1-dependent DNA repair activity. This finding suggests that pulmonary fibrosis may develop and worsen due to the presence of apoptosis-resistant lung fibroblasts in cisplatin-treated cancer patients.