Merck
  • The different expression of tumor suppressors, RASSF1A, RUNX3, and GSTP1, in patients with alcoholic steatohepatitis (ASH) vs non-alcoholic steatohepatitis (NASH).
The different expression of tumor suppressors, RASSF1A, RUNX3, and GSTP1, in patients with alcoholic steatohepatitis (ASH) vs non-alcoholic steatohepatitis (NASH).
Experimental and molecular pathology (2019-04-06)
Yue Jia, Ping Ji, Barbara French, Brittany Tillman, Samuel W French
ABSTRACT
As the fifth most common cancer and the second leading cause of cancer related deaths worldwide, hepatocellular carcinoma (HCC) causes up to one million deaths annually. Alcoholic steatohepatitis (ASH) and non-alcoholic steatohepatitis (NASH) are becoming the two major risk factors because both may develop liver fibrosis and hepatocellular carcinoma (HCC) if left untreated. However, compared with 3-10% of patients with ASH may progress to HCC annually, about only 0.5% NASH patients may progress to HCC annually. The present study is to clarify the protein expression differences of tumor suppressor genes (TSGs) between ASH and NASH. In liver biopsied specimens from NASH and ASH patients, using an immunofluorescence method and morphometrically quantitating the fluorescence intensity, we studied the protein expression within hepatocytes cytoplasm of candidate TSGs including RUNX3, GSTP1, and RASSF1A. Compared with the control group of patients, the expression levels of all three proteins were upregulated in the ASH group of patients (p < .001 in all molecules). While RUNX3 was upregulated, GSTP1 and RASSF1 did not change in the NASH group of patients. The most important finding is that compared with the ASH group of patients, the expression levels of all three TSG proteins, RUNX3, GSTP1, and RASSF1, were significantly lower in the NASH group of patients (p < .001 in all three molecules). These results confirmed our previous finding that there are significant differences of many molecules including TSGs that changed in NASH compared to ASH. Thus, we conclude that there are significantly different TSGs and pathways involved during the pathogenesis of HCC development in NASH compared to ASH that may help to develop different strategies for prevention and treatment of NASH and ASH patients.