Merck
  • FBXL3 is regulated by miRNA-4735-3p and suppresses cell proliferation and migration in non-small cell lung cancer.
FBXL3 is regulated by miRNA-4735-3p and suppresses cell proliferation and migration in non-small cell lung cancer.
Pathology, research and practice (2018-12-31)
Dazhong Wang, Xin Han, Chan Li, Weijun Bai
ABSTRACT
Non-small cell lung cancer (NSCLC) is the most common type of primary lung cancer and regarded as cancer killer. The aim of this study was to discover the detailed function and molecular mechanism of F-box and leucine rich repeat protein 3 (FBXL3) in NSCLC. In this study, the expression level of FBXL3 in NSCLC tissues and cell lines was firstly examined and identified. Moreover, the relationship between FBXL3 and the overall survival rate of NSCLC patients was analyzed by Kaplan-Meier survival curve. Functionally, MTT, colony formation assay and transwell assays were performed to determine the role of FBXL3 in regulating NSCLC cell proliferation, migration and invasion. The proliferation and migration were suppressed by overexpression of FBXL3, indicating the potential tumor suppressive role of FBXL3 in NSCLC. In addition, the dual-luciferase reporter and RNA pull-down assays revealed that miR-4735-3p was a novel upstream modulator of FBXL3. Further study showed that miR-4735-3p was upregulated in NSCLC tissues and cell lines. Finally, rescue assays and function assays revealed that miR-4735-3p exerted oncogenic function in NSCLC, and this function can be attenuated by FBXL3. Taken together, FBXL3 was regulated by miR-4735-3p and suppressed cell proliferation and invasion in non-small cell lung cancer.