Merck
Design of selective Cathepsin inhibitors.
Bioorganic & medicinal chemistry letters (2009-07-21)
Paul A Bethel, Stefan Gerhardt, Emma V Jones, Peter W Kenny, Galith I Karoutchi, Andrew D Morley, Keith Oldham, Neil Rankine, Martin Augustin, Stephan Krapp, Hannes Simader, Stefan Steinbacher
ABSTRACT
A number of molecular recognition features have been exploited in structure-based design of selective Cathepsin inhibitors.
Sigma-Aldrich
1-Methyl-1H-pyrazole-5-boronic acid pinacol ester, 97%
Sigma-Aldrich
1-Boc-pyrazole-4-boronic acid pinacol ester, 97%