Merck
  • Highly Efficient Nanocarbon Coating Layer on the Nanostructured Copper Sulfide-Metal Organic Framework Derived Carbon for Advanced Sodium-Ion Battery Anode.
Highly Efficient Nanocarbon Coating Layer on the Nanostructured Copper Sulfide-Metal Organic Framework Derived Carbon for Advanced Sodium-Ion Battery Anode.
Materials (Basel, Switzerland) (2019-04-26)
Chiwon Kang, Yongwoo Lee, Ilhwan Kim, Seungmin Hyun, Tae Hoon Lee, Soyeong Yun, Won-Sub Yoon, Youngkwang Moon, Jinkee Lee, Sunkook Kim, Hoo-Jeong Lee
ABSTRACT
High theoretical capacity and low-cost copper sulfide (CuxS)-based anodes have gained great attention for advanced sodium-ion batteries (SIBs). However, their practical application may be hindered due to their unstable cycling performance and problems with the dissolution of sodium sulfides (NaxS) into electrolyte. Here, we employed metal organic framework (MOF-199) as a sacrificial template to fabricate nanoporous CuxS with a large surface area embedded in the MOF-derived carbon network (CuxS-C) through a two-step process of sulfurization and carbonization via H2S gas-assisted plasma-enhanced chemical vapor deposition (PECVD) processing. Subsequently, we uniformly coated a nanocarbon layer on the Cu1.8S-C through hydrothermal and subsequent annealing processes. The physico-chemical properties of the nanocarbon layer were revealed by the analytical techniques of high-resolution transmission electron microscopy (HRTEM), energy-dispersive X-ray spectroscopy (EDS), and scanning electron microscopy (SEM). We acquired a higher SIB performance (capacity retention (~93%) with a specific capacity of 372 mAh/g over 110 cycles) of the nanoporous Cu1.8S-C/C core/shell anode materials than that of pure Cu1.8S-C. This encouraging SIB performance is attributed to the key roles of a nanocarbon layer coated on the Cu1.8S-C to accommodate the volume variation of the Cu1.8S-C anode structure during cycling, enhance electrical conductivity and prevent the dissolution of NaxS into the electrolyte. With these physico-chemical and electrochemical properties, we ensure that the Cu1.8S-C/C structure will be a promising anode material for large-scale and advanced SIBs.