Merck
  • Rh2(R-TPCP)4-catalyzed enantioselective [3+2]-cycloaddition between nitrones and vinyldiazoacetates.
Rh2(R-TPCP)4-catalyzed enantioselective [3+2]-cycloaddition between nitrones and vinyldiazoacetates.
Journal of the American Chemical Society (2013-09-13)
Changming Qin, Huw M L Davies
ABSTRACT
Rhodium-catalyzed reaction of vinyldiazoacetates with nitrones results in a formal [3+2]-cycloaddition to generate 2,5-dihydroisoxazoles with high levels of asymmetric induction. The cascade reaction begins with a vinylogous addition event, followed by an iminium addition ring-closure/hydride migration/alkene isomerization cascade. Dirhodium tetrakis(triarylcyclopropanecarboxylates) are the optimum catalysts for this process.
Sigma-Aldrich
Rh2(R-p-Ph-TPCP)4