Merck
  • A New Conjugation Method Used for the Development of an Immunoassay for the Detection of Amanitin, a Deadly Mushroom Toxin.
A New Conjugation Method Used for the Development of an Immunoassay for the Detection of Amanitin, a Deadly Mushroom Toxin.
Toxins (2018-07-01)
Candace S Bever, Bogdan Barnych, Robert Hnasko, Luisa W Cheng, Larry H Stanker
ABSTRACT
One of the deadliest mushrooms is the death cap mushroom, Amanita phalloides. The most toxic constituent is α-amanitin, a bicyclic octapeptide, which damages the liver and kidneys. To develop a new tool for detecting this toxin, polyclonal antibodies were generated and characterized. Both α- and β-amanitin were coupled to carrier proteins through four different linking chemistries, one of which has never before been described. These conjugates were evaluated for their effectiveness in generating antibodies specific for the free toxin, as well as their utility in formatting heterogeneous assays with high sensitivity. Ultimately, these efforts yielded a newly described conjugation procedure utilizing periodate oxidation followed by reductive amination that successfully resulted in generating sensitive immunoassays (limit of detection (LOD), ~1.0 µg/L). The assays were characterized for their selectivity and were found to equally detect α-, β-, and γ-amanitin, and not cross-react with other toxins tested. Toxin detection in mushrooms was possible using a simple sample preparation method. This enzyme-linked immunosorbent assay (ELISA) is a simple and fast test, and readily detects amatoxins extracted from A. phalloides.
Product Description
Supelco
Microcystin-LR solution, 10 μg/mL in methanol, analytical standard