Merck
  • Improved LC/MS Methods for the Analysis of Metal-Sensitive Analytes Using Medronic Acid as a Mobile Phase Additive.
Improved LC/MS Methods for the Analysis of Metal-Sensitive Analytes Using Medronic Acid as a Mobile Phase Additive.
Analytical chemistry (2018-07-07)
Jordy J Hsiao, Oscar G Potter, Te-Wei Chu, Hongfeng Yin
ABSTRACT
Phosphorylated compounds and organic acids with multiple carboxylate groups are commonly observed to have poor peak shapes and signal in LC/MS experiments. The poor peak shape is caused by the presence of trace metals, particularly iron, contributed from a variety of sources within the chromatographic system. To ameliorate this problem, different solvent additives were investigated to reduce the amount of metal in the flow path to achieve better analytical performance for these metal-sensitive compounds. Here, we introduce the use of a solvent additive that can significantly improve the peak shapes and signal of metal-sensitive metabolites for LC/MS analysis. Moreover, the additive is shown to be amenable for other metal-sensitive applications, such as the analysis of phosphopeptides and polar phosphorylated pesticides, where the instruments could be used in either positive or negative analysis mode.
Sigma-Aldrich
MS PhosphoMix 1 Light, Phosphopeptide Standard for MS
Pricing and availability is not currently available.
Sigma-Aldrich
MS PhosphoMix 3 Light, Phosphopeptide Standard for MS
Pricing and availability is not currently available.