Merck
  • p21waf1/cip1 is down-regulated in conjunction with up-regulation of c-Fos in the lymphocytes of rheumatoid arthritis patients.
p21waf1/cip1 is down-regulated in conjunction with up-regulation of c-Fos in the lymphocytes of rheumatoid arthritis patients.
Biochemical and biophysical research communications (2003-04-23)
Mari Hikasa, Eri Yamamoto, Hiroki Kawasaki, Koichiro Komai, Kazuko Shiozawa, Akira Hashiramoto, Yasushi Miura, Shunichi Shiozawa
ABSTRACT
Features characteristic to rheumatoid arthritis (RA) including synovial overgrowth and joint destruction are experimentally produced by augmenting c-fos gene expression. We show that cyclin dependent kinase inhibitor p21waf1/cip1, that inhibits cell proliferation, is down-regulated in conjunction with up-regulation of c-fos in the lymphocytes of patients with RA. As to the mechanism of down-regulation of p21waf1/cip1 gene expression, transfection studies in U937 cells showed that c-fos down-regulated phosphorylation and dimerization of signal transducers and activators of transcription (STAT) 1, thereby inhibiting interferon -induced transactivation of p21waf1/cip1. Phosphorylation of STAT1 was indeed decreased in the lymphocytes of patients with RA. Thus, under overexpression of c-fos gene, c-Fos inactivates STAT1 to down-regulate p21waf1/cip1 gene expression in the lymphocytes of patients with RA, and in this way may enhance proliferation of lymphocytes.
Sigma-Aldrich
FOS, GST tagged human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution