Merck
  • Production of d-Branched-Chain Amino Acids by Lactic Acid Bacteria Carrying Homologs to Isoleucine 2-Epimerase of Lactobacillus buchneri.
Production of d-Branched-Chain Amino Acids by Lactic Acid Bacteria Carrying Homologs to Isoleucine 2-Epimerase of Lactobacillus buchneri.
Frontiers in microbiology (2018-07-31)
Yuta Mutaguchi, Kano Kasuga, Ikuo Kojima
ABSTRACT
Isoleucine 2-epimerase (ILEP) is a novel branched-chain amino acid racemase isolated from Lactobacillus buchneri. In this study, we examined production of free d-branched-chain amino acids such as d-valine, d-leucine, and d-allo-isoleucine, using lactic acid bacteria carrying homologs to ILEP. Twelve selected strains of lactic acid bacteria were grown at optimal growth temperatures and accumulation of d-branched-chain amino acids in the medium was monitored in exponential, early stationary, and stationary phases. To analyze the d-branched-chain amino acids, enantiomers in the medium were initially converted into diastereomers using pre-column derivatization with o-phthaldialdehyde plus N-isobutyryl-l-cysteine. The resultant fluorescent isoindole derivatives were analyzed on an octadecylsilyl stationary phase using ultra-high performance liquid chromatography. The analyses revealed that the seven following lactic acid bacteria carrying homologs showing 53-60% amino acid sequence identity to the L. buchneri ILEP accumulate d-branched-chain amino acids: Lactobacillus fermentum and Weissella paramesenteroides produce d-valine, d-leucine, and d-allo-isoleucine; Lactobacillus reuteri, Leuconostoc mesenteroides subsp. mesenteroides, and Leuconostoc gelidum subsp. gasicomitatum accumulate d-leucine and d-allo-isoleucine; and Lactobacillus vaginalis and Leuconostoc pseudomesenteroides produce d-allo-isoleucine. These results suggest that d-branched-chain amino acids are produced by a variety of lactic acid bacteria species, particularly those carrying homologs to the ILEP.
Product Description
Supelco
N-Isobutyryl-L-cysteine, for chiral derivatization, LiChropur, ≥97.0%