Merck
  • Related RNA polymerase-binding regions in human RAP30/74 and Escherichia coli sigma 70.
Related RNA polymerase-binding regions in human RAP30/74 and Escherichia coli sigma 70.
Science (New York, N.Y.) (1991-08-23)
S McCracken, J Greenblatt
ABSTRACT
RAP30/74 is a heteromeric general transcription initiation factor that binds to mammalian RNA polymerase II. The RAP30 subunit contains a region that is similar in amino acid sequence to the RNA polymerase-binding domain of the Escherichia coli transcription initiation factor sigma 70 (sigma 70). Mammalian RNA polymerase II specifically protected a serine residue in the sigma 70-related region of RAP30 from phosphorylation in vitro. In addition, human RAP30/74 bound to Escherichia coli RNA polymerase and was displaced by sigma 70. These results suggest that RAP30 and sigma 70 have functionally related RNA polymerase-binding regions.
Sigma-Aldrich
TFIIF (RAP30 subunit) human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
TFIIF, Rap30 subunit, GST tagged human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
TFIIF (RAP30+Rap74) human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)