Skip to Content
Merck
vibrant-m

Valsartan Related Compound E

vibrant-m
Synonym(s):
N-Valeryl-N-{[2′-(1H-tetrazole-5-yl)biphenyl-4-yl]methyl}-L-valine methyl ester, Valsartan methyl ester
Empirical Formula (Hill Notation):
C25H31N5O3
CAS Number:
Molecular Weight:
449.55

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1