![Valproic Acid Impurity H Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/225/525/48a00246-21e9-49d5-8436-b0d2eb7844f6/640/48a00246-21e9-49d5-8436-b0d2eb7844f6.png)
Valproic Acid Impurity H
![Valproic Acid Impurity H Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/225/525/48a00246-21e9-49d5-8436-b0d2eb7844f6/640/48a00246-21e9-49d5-8436-b0d2eb7844f6.png)
Synonym(s):
Valeronitrile, Butyl cyanide, Pentanenitrile
Linear Formula:
CH3(CH2)3CN
CAS Number:
Molecular Weight:
83.13
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing