trans-p-Methoxycinnamic acid ethyl ester
Synonym(s):
Ethyl trans-4-methoxycinnamate, (E)-3-(4-Methoxyphenyl)-2-propenoic acid ethyl ester, trans-4-Methoxycinnamic acid ethyl ester, Ethyl (E)-3-(4-methoxyphenyl)-2-propenoate
Empirical Formula (Hill Notation):
C12H14O3
CAS Number:
Molecular Weight:
206.24
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing