Skip to Content
Merck
Terbinafine for system suitability European Pharmacopoeia (EP) Reference Standard

Terbinafine for system suitability

Terbinafine for system suitability European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Terbinafine hydrochloride, trans-N-(6,6-Dimethyl-2-hepten-4-ylyl)-N-methyl-1-naphthylmethylamine hydrochloride
Empirical Formula (Hill Notation):
C21H25N · HCl
CAS Number:
Molecular Weight:
327.89

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1