SUPELCOGEL™ 8H (9 µm) HPLC Columns
Synonym(s):
SUPELCOGEL™ 8% Crosslinked HPLC Column, SUPELCOGEL™ 8% Crosslinked Guard Column
Products
Product Category
Brand
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Column Material
Package Size
Particle Size (µm)
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing