
Spirotetramat Metabolite BYI08330-cis-enol

Synonym(s):
cis-3-(2,5-Dimethylphenyl)-4-hydroxy-8-methoxy-1-azaspiro[4.5]dec-3-en-2-one
Empirical Formula (Hill Notation):
C18H23NO3
CAS Number:
Molecular Weight:
301.38
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing