
Salmonella ChromoSelect Agar

Synonym(s):
Salmonella Agar ChromoSelect
Products
Product Category
Brand
pH Value
Physical Form
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing