![Safinamide Mesylate Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/300/388/772363b0-dc52-42ec-801a-27ee64119c86/640/772363b0-dc52-42ec-801a-27ee64119c86.png)
Safinamide Mesylate
![Safinamide Mesylate Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/300/388/772363b0-dc52-42ec-801a-27ee64119c86/640/772363b0-dc52-42ec-801a-27ee64119c86.png)
Synonym(s):
Safinamide mesylate salt, (S)-(+)-2-[[4-(3-Fluorobenzoxy)benzyl]amino]propanamide; (S)-2-[[4-[(3-Fluorobenzyl)oxy]benzyl]amino]propanamide; FCE-26743; PNU-151774E, NW-1015;
Empirical Formula (Hill Notation):
C17H19FN2O2 · xCH4O3S
CAS Number:
Molecular Weight:
302.34 (free base basis)
Products
Brand
Formula Weight
Gene Alias
Physical Form
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing