(R)-Amlodipine
Synonym(s):
(R)-Amlodipine
Empirical Formula (Hill Notation):
C20H25ClN2O5
CAS Number:
Molecular Weight:
408.88
Products
Formula Weight
Manufacturer
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing