
Paricalcitol

Synonym(s):
(1α, 3β, 7E,22E)-19-Nor-9,10-secoergosta-5,7,22-triene-1,3,25-triol, (1R,3R)-5-((E)-2-((1R,3aS,7aR)-1-((2R,5S,E)-6-hydroxy-5,6-dimethylhept-3-en-2-yl)-7a-methyldihydro-1H-inden-4(2H,5H,6H,7H,7aH)-ylidene)ethylidene)cyclohexane-1,3-diol, (1alpha.3beta,7E,22E)-19-Nor-9,10-secoergosta-5,7,22-triene-1,3,25-triol, (7E,22E)-19-Nor-9,10-secoergosta-5,7,22-triene-1alpha,3beta,25-triol, 19-Nor-1alpha,25-dihydroxyvitamin D2
Empirical Formula (Hill Notation):
C27H44O3
CAS Number:
Molecular Weight:
416.64
Products
Brand
Color
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing