Skip to Content
Merck
Nicardipine for system suitability European Pharmacopoeia (EP) Reference Standard

Nicardipine for system suitability

Nicardipine for system suitability European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Nicardipine hydrochloride, 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid methyl 2-[methyl(phenylmethyl)amino]ethyl ester hydrochloride, YC-93
Empirical Formula (Hill Notation):
C26H29N3O6 · HCl
CAS Number:
Molecular Weight:
515.99
EC Number:

Products

Formula Weight
Gene Alias
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1