Moxidectin for system suitability
Synonym(s):
Moxidectin
Empirical Formula (Hill Notation):
C37H53NO8
CAS Number:
Molecular Weight:
639.82
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing