![Monoclonal Anti-HIST1H3A antibody produced in mouse Prestige Antibodies® Powered by Atlas Antibodies, clone CL4968, purified immunoglobulin, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/221/880/c1f77b15-fbce-4c76-8855-8ebe06fd72c8/640/c1f77b15-fbce-4c76-8855-8ebe06fd72c8.jpg)
Monoclonal Anti-HIST1H3A antibody produced in mouse
![Monoclonal Anti-HIST1H3A antibody produced in mouse Prestige Antibodies® Powered by Atlas Antibodies, clone CL4968, purified immunoglobulin, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/221/880/c1f77b15-fbce-4c76-8855-8ebe06fd72c8/640/c1f77b15-fbce-4c76-8855-8ebe06fd72c8.jpg)
Synonym(s):
H3, H3FA, H3FB, H3FC, H3FD, H3FF, H3FHH3FI, H3FJ, H3FK, H3FL
Products
Product Category
Brand
Clonality
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing