
Luteolin 7-O-Glucoside

Synonym(s):
Luteolin 7-O-β-D-glucoside, 3′,4′,5,7-Tetrahydroxyflavone 7-glucoside, Cynaroside, Glucoluteolin, Glucosylluteolin, Luteoloside
Empirical Formula (Hill Notation):
C21H20O11
CAS Number:
Molecular Weight:
448.38
Products
Formula Weight
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing