
Glyoxalbis(2-hydroxyanil)

Synonym(s):
Glyoxalbis(2-hydroxyanil)
Empirical Formula (Hill Notation):
C14H12N2O2
CAS Number:
Molecular Weight:
240.26
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing