Skip to Content
Merck
Fluo 3-AM ≥90% (TLC)

Fluo 3-AM

Fluo 3-AM ≥90% (TLC)
Synonym(s):
4-(6-Acetoxymethoxy-2,7-dichloro-3-oxo-9-xanthenyl)-4′-methyl-2,2′(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, FLUO 3/AM
Empirical Formula (Hill Notation):
C51H50Cl2N2O23
CAS Number:
Molecular Weight:
1129.85

Products

Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment