Skip to Content
Merck
Endothelin 1 ≥97% (HPLC), powder

Endothelin 1

Endothelin 1 ≥97% (HPLC), powder
Synonym(s):
ET-1
Empirical Formula (Hill Notation):
C109H159N25O32S5
CAS Number:
Molecular Weight:
2491.90

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing