Copper tetramibi tetrafluoroborate
Synonym(s):
Tetrakis(2-methoxyisobutyl isonitrile)copper(I) tetrafluoroborate, (2-Methoxy-2-methylpropaneisocyanide)copper(I) tetrafluoroborate, MIBI copper complex
Empirical Formula (Hill Notation):
C24H44BCuF4N4O4
CAS Number:
Molecular Weight:
602.98
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing