Skip to Content
Merck
Cobalt carbonyl moistened with hexane (hexane 1-10%), ≥90% (Co)

Cobalt carbonyl

Cobalt carbonyl moistened with hexane (hexane 1-10%), ≥90% (Co)
Synonym(s):
Dicobalt octacarbonyl
Linear Formula:
Co2(CO)8
CAS Number:
Molecular Weight:
341.95

Products

Brand
Formula Weight
Physical Form
Purity
Reagent Type
Core
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing