
Cinobufagin

Synonym(s):
14,15β-Epoxy-3β,16β-dihydroxy-5β,20(22)-bufadienolide 16-acetate, 5β,20(22)-Bufadienolide-3β,16β-diol-14,15β-epoxy 16-acetate
Empirical Formula (Hill Notation):
C26H34O6
CAS Number:
Molecular Weight:
442.54
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing