
Boc-7-Ahp-OH

Synonym(s):
7-(Boc-amino)enanthic acid, 7-(Boc-amino)heptanoic acid, Boc-7-aminoheptanoic acid
Empirical Formula (Hill Notation):
C12H23NO4
CAS Number:
Molecular Weight:
245.32
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing