Skip to Content
Merck
Benzaldehyde dimethyl acetal ≥98%, FG

Benzaldehyde dimethyl acetal

Benzaldehyde dimethyl acetal ≥98%, FG
Synonym(s):
α,α-Dimethoxytoluene
Linear Formula:
C6H5CH(OCH3)2
CAS Number:
Molecular Weight:
152.19
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing