![Asperosaponin VI phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/278/832/1f730e53-1ba7-46fa-a507-ca0f920996ac/640/1f730e53-1ba7-46fa-a507-ca0f920996ac.png)
Asperosaponin VI
![Asperosaponin VI phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/278/832/1f730e53-1ba7-46fa-a507-ca0f920996ac/640/1f730e53-1ba7-46fa-a507-ca0f920996ac.png)
Synonym(s):
Akebia saponin D, 3-O-α-L-arabinopyranosyl hederagenin-28-β-D-glucopyranoside-(1→6)-β-D-glucopyranoside, ASD, Asperosaponin VI, Leiyemudanoside A, Tauroside St-G0-1
Empirical Formula (Hill Notation):
C47H76O18
CAS Number:
Molecular Weight:
929.10
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing