
Anti-Mouse IgG (whole molecule) F(ab′)2 fragment–Alkaline Phosphatase antibody produced in sheep

Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1