Skip to Content
Merck
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal clone 1A4, purified from hybridoma cell culture

Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal

Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal clone 1A4, purified from hybridoma cell culture
Synonym(s):
Monoclonal Anti-Actin, α-Smooth Muscle, SMA

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Application
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1