Aminoglutethimide impurity A
Synonym(s):
(3RS)-3-(3-Aminophenyl)-3-ethylpiperidine-2,6-dione, 3-Aminoglutethimide
Empirical Formula (Hill Notation):
C13H16N2O2
CAS Number:
Molecular Weight:
232.28
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing