![α-Amylcinnamaldehyde dimethyl acetal United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/184/544/47256f4f-f0ae-48de-ba03-eb9c22e918ea/640/47256f4f-f0ae-48de-ba03-eb9c22e918ea.png)
α-Amylcinnamaldehyde dimethyl acetal
![α-Amylcinnamaldehyde dimethyl acetal United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/184/544/47256f4f-f0ae-48de-ba03-eb9c22e918ea/640/47256f4f-f0ae-48de-ba03-eb9c22e918ea.png)
Synonym(s):
α-Pentylcinnamaldehyde dimethyl acetal, [2-(Dimethoxymethyl)-1-hepten-1-yl]benzene
Linear Formula:
C6H5CH=C[(CH2)4CH3]CH(OCH3)2
CAS Number:
Molecular Weight:
248.36
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing