Skip to Content
Merck
3,4-Dichlorobenzyl alcohol for synthesis

3,4-Dichlorobenzyl alcohol

3,4-Dichlorobenzyl alcohol for synthesis
Synonym(s):
3,4-Dichlorobenzyl alcohol
Empirical Formula (Hill Notation):
C7H6Cl2O
CAS Number:
Molecular Weight:
177.03

Products

Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing